Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Diiodoperfluorobutane ≥97%

Supplier: MATRIX SCIENTIFIC MS

004915-25G 004915-5G
101777-852EA 361.54 USD InStock
101777-852 101777-850
1,4-Diiodoperfluorobutane ≥97%
1,4-Diiodoperfluorobutane
Hazard: Toxic


Formula: C₄F₈I₂
Boiling Pt: 85 °C (100 mmHg)
MDL Number: MFCD00042263
CAS Number: 375-50-8
Order Now
Specification Test Results
Boiling point 85°/100mm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty