Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,6-Difluoro-3-methylphenyl)acetonitrile

Supplier: MATRIX SCIENTIFIC MS

005022-1G 005022-5G
101777-908EA 104.62 USD InStock
101777-908 101777-910
(2,6-Difluoro-3-methylphenyl)acetonitrile
(2,6-Difluoro-3-methylphenyl)acetonitrile
Hazard: Irritant-Harmful


Formula: C₉H₇F₂N
MDL Number: MFCD01631337
CAS Number: 261944-04-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty