Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-4-(trifluoromethyl)aniline ≥99%

Supplier: MATRIX SCIENTIFIC MS

005056-25G 005056-100G
101777-920EA 35.38 USD InStock
101777-920 101777-922
2,6-Dichloro-4-(trifluoromethyl)aniline ≥99%
2,6-Dichloro-4-(trifluoromethyl)aniline
Hazard: Toxic


Formula: C₇H₄Cl₂F₃N
Boiling Pt: 60-62°C/1mm
Melting Pt: 35-37°C
Density: 1.532
MDL Number: MFCD00052918
CAS Number: 24279-39-8
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Boiling point 60-62°/1mm
Density 1.532

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty