Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-p-cresol

Supplier: MATRIX SCIENTIFIC MS

005078-5G 005078-1G
101777-938EA 52.31 USD InStock
101777-938 101777-936
2,3-Difluoro-p-cresol
2,3-Difluoro-p-cresol
Hazard: Corrosive


Formula: C₇H₆F₂O
Melting Pt: 35…38 °C
MDL Number: MFCD01631651
CAS Number: 261763-45-5
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty