Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-6-(trifluoromethyl)aniline ≥97%

Supplier: MATRIX SCIENTIFIC MS

005122-1G 005122-5G
101777-956EA 80 USD InStock
101777-956 101777-958
2,4-Dichloro-6-(trifluoromethyl)aniline ≥97%
2,4-Dichloro-6-(trifluoromethyl)aniline
Hazard: Irritant-Harmful


Formula: C₇H₄Cl₂F₃N
MW: 230.02
Boiling Pt: 49…52 °C (0.4 mmHg)
MDL Number: MFCD00174313
CAS Number: 62593-17-3
UN: 3082
ADR: 9 III
Order Now
Specification Test Results
Boiling point 49-52°/0.4

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty