Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoroaniline ≥98%

Supplier: MATRIX SCIENTIFIC MS

005149-100G 005149-5G
101777-998EA 598.46 USD InStock
101777-998 101777-994
2,3-Difluoroaniline ≥98%
2,3-Difluoroaniline
Hazard: Toxic


Formula: C₆H₅F₂N
Boiling Pt: 68…69 °C (12 mmHg)
Density: 1.274
MDL Number: MFCD00010298
CAS Number: 4519-40-8
UN: 2941
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 68-69°/12m
Density 1.274
ND20 1.514

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty