Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluoroanisole ≥98%

Supplier: MATRIX SCIENTIFIC MS

005153-5G 005153-25G
101778-012EA 13.85 USD InStock
101778-012 101778-014
3,4-Difluoroanisole ≥98%
3,4-Difluoroanisole
Hazard: Flammable


Formula: C₇H₆F₂O
MW: 144.12
Boiling Pt: 139
Melting Pt: -30
Density: 1.216
MDL Number: MFCD00010730
CAS Number: 115144-40-6
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Density 1.216
ND20 1.469

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty