Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichlorobenzenesulfonyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

005172-5G 005172-25G
101778-034EA 21.54 USD InStock
101778-034 101778-036
2,4-Dichlorobenzenesulfonyl chloride ≥97%
2,4-Dichlorobenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₆H₃Cl₃O₂S
Melting Pt: 54…56 °C
MDL Number: MFCD00052712
CAS Number: 16271-33-3
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Melting point 54-56°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty