Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorobenzenesulfonyl chloride ≥98%

Supplier: MATRIX SCIENTIFIC MS

005185-25G 005185-100G
101778-070EA 158.46 USD InStock
101778-070 101778-072
3,4-Dichlorobenzenesulfonyl chloride ≥98%
3,4-Dichlorobenzenesulphonyl chloride
TCA listed: T
Hazard: Corrosive/Moisture Sensitive


Formula: C₆H₃Cl₃O₂S
Boiling Pt: 158…160 °C (15 mmHg)
Melting Pt: 22 °C
Density: 1.572
MDL Number: MFCD00041255
CAS Number: 98-31-7
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Boiling point 158-160°/1
Density 1.572
ND20 1.589

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty