Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorobenzyl bromide ≥98%

Supplier: MATRIX SCIENTIFIC MS

005236-100G 005236-5G 005236-25G
101778-134EA 746.15 USD InStock
101778-134 101778-130 101778-132
2,3-Difluorobenzyl bromide ≥98%
2,3-Difluorobenzyl bromide
Hazard: Corrosive/Refrigerate/Lachrymator


Formula: C₇H₅BrF₂
Density: 1.628
MDL Number: MFCD00042488
CAS Number: 113211-94-2
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Density 1.628
ND20 1.528

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty