Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl chlorofluoroacetate ≥97%

Supplier: MATRIX SCIENTIFIC MS

005277-25G 005277-100G
101778-204EA 130.77 USD InStock
101778-204 101778-206
Ethyl chlorofluoroacetate ≥97%
Ethyl chlorofluoroacetate
TCA listed: T
Hazard: Flammable/Lachrymator


Formula: C₄H₆ClFO₂
Boiling Pt: 128-130°C
Density: 1.225
MDL Number: MFCD00039333
CAS Number: 401-56-9
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Boiling point 128-130°
Density 1.225
ND20 1.3927

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty