Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o,α,α,α-Tetrafluoroanisole

Supplier: MATRIX SCIENTIFIC MS

005327-5G 005327-25G
101778-262EA 21.54 USD InStock
101778-262 101778-264
o,α,α,α-Tetrafluoroanisole
o,α,α,α-Tetrafluoroanisole
Hazard: Flammable


Formula: C₇H₄F₄O
MW: 180.1
Boiling Pt: 105…106 °C
MDL Number: MFCD00236322
CAS Number: 2106-18-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty