Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethoxy)benzonitrile ≥97%

Supplier: MATRIX SCIENTIFIC MS

005367-25G 005367-5G
101778-310EA 50.77 USD InStock
101778-310 101778-308
2-(Trifluoromethoxy)benzonitrile ≥97%
2-(Trifluoromethoxy)benzonitrile
Hazard: Toxic/Irritant


Formula: C₈H₄F₃NO
Boiling Pt: 80…82 °C (15 mmHg)
MDL Number: MFCD00042409
CAS Number: 63968-85-4
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 80-82°/15m
ND20 1.449

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty