Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)benzylamine ≥98%

Supplier: MATRIX SCIENTIFIC MS

005394-5G 005394-25G
101778-324EA 24.62 USD InStock
101778-324 101778-326
2-(Trifluoromethyl)benzylamine ≥98%
2-(Trifluoromethyl)benzylamine
Hazard: Corrosive


Formula: C₈H₈F₃N
MW: 175.15
Boiling Pt: 108
Density: 1.249
MDL Number: MFCD00010297
CAS Number: 3048-01-9
Order Now
Specification Test Results
Density 1.249
ND20 1.471

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty