Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)pyridine ≥97%

Supplier: MATRIX SCIENTIFIC MS

005457-5G 005457-25G
101778-344EA 58.46 USD InStock
101778-344 101778-346
3-(Trifluoromethyl)pyridine ≥97%
3-(Trifluoromethyl)pyridine
Hazard: Flammable/Irritant


Formula: C₆H₄F₃N
Boiling Pt: 116…117 °C
MDL Number: MFCD00100032
CAS Number: 3796-23-4
UN: 1992
ADR: 3,III
Order Now
Specification Test Results
Boiling point 116-117°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty