Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoropyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS

005610-25G 005610-100G
101778-534EA 30.77 USD InStock
101778-534 101778-536
2,6-Difluoropyridine ≥98%
2,6-Difluoropyridine
Hazard: Flammable


Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 124.5 °C (743 mmHg)
Density: 1.268
MDL Number: MFCD00006227
CAS Number: 1513-65-1
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Boiling point 124.5°/743
Density 1.268
ND20 1.437

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty