Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptafluorobutyryl chloride ≥98%

Supplier: MATRIX SCIENTIFIC MS

005975-25G 005975-100G
101778-784EA 323.08 USD InStock
101778-784 101778-786
Heptafluorobutyryl chloride ≥98%
Heptafluorobutyryl chloride
Hazard: Corrosive/Lachrymator


Formula: C₄ClF₇O
Boiling Pt: 38…39 °C
Density: 1.556
MDL Number: MFCD00000707
CAS Number: 375-16-6
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Boiling point 38-39°
Density 1.556
ND20 1.288

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty