Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Difluoromethoxy)aniline ≥97%

Supplier: MATRIX SCIENTIFIC MS

006024-5G 006024-25G
101778-826EA 15.38 USD InStock
101778-826 101778-828
4-(Difluoromethoxy)aniline ≥97%
4-(Difluoromethoxy)aniline
Hazard: Toxic/Irritant-Harmful


Formula: C₇H₇F₂NO
Boiling Pt: 98-100°C/5mm
Density: 1.283
MDL Number: MFCD00085005
CAS Number: 22236-10-8
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 98-100°/5m
Density 1.283
ND20 1.505

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty