Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluoropropionyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

006133-25G 006133-100G
101778-894EA 592.31 USD InStock
101778-894 101778-896
Pentafluoropropionyl chloride ≥97%
Pentafluoropropionyl chloride
TCA listed: T
Hazard: Toxic by inhalation/Corrosive/Nonflammable Gas


Formula: C₃ClF₅O
Boiling Pt: 7…9 °C
MDL Number: MFCD00039308
CAS Number: 422-59-3
Order Now
Specification Test Results
Boiling point 7-9°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty