Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsilyl trifluoroacetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

006197-100G 006197-25G
101778-936EA 243.08 USD InStock
101778-936 101778-934
Trimethylsilyl trifluoroacetate ≥95%
Trimethylsilyl trifluoroacetate
TCA listed: T
Hazard: Flammable


Formula: C₅H₉F₃O₂Si
MW: 186.21 g/mol
Boiling Pt: 88…90 °C
Density: 1.078
MDL Number: MFCD00000413
CAS Number: 400-53-3
Order Now
Specification Test Results
Boiling point 88-90°
Density 1.078
ND20 1.338

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty