Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroethyl isothiocyanate ≥99%

Supplier: MATRIX SCIENTIFIC MS

006327-5G 006327-25G
101779-022EA 66.15 USD InStock
101779-022 101779-024
2-Chloroethyl isothiocyanate ≥99%
2-Chloroethyl isothiocyanate
Hazard: Flammable/Toxic/Moisture Sensitive


Formula: C₃H₄ClNS
MDL Number: MFCD00041125
CAS Number: 6099-88-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty