Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-Methylstyrene (cis- and trans- mixture) racemate

Supplier: MATRIX SCIENTIFIC MS

006410-5G 006410-25G
101779-058EA 143.08 USD InStock
101779-058 101779-060
β-Methylstyrene (cis- and trans- mixture) racemate
beta-Methylstyrene (cis- and trans- mixture)
Hazard: Flammable/Refrigerate/Lachrymator


Formula: C₉H₁₀
Boiling Pt: 172…173 °C
Density: 0.91
MDL Number: MFCD00009280
CAS Number: 637-50-3
Order Now
Specification Test Results
Boiling point 172-173°
Density 0.91

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty