Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Fluorophenylacetic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

006715-5G 006715-1G
101779-232EA 332.31 USD InStock
101779-232 101779-230
α-Fluorophenylacetic acid ≥98%
α-Fluorophenylacetic acid
Hazard: Toxic


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 81…82 °C
MDL Number: MFCD00004221
CAS Number: 1578-63-8
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 81-82°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty