Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: MATRIX SCIENTIFIC MS

006783-100G 006783-25G
101779-284EA 89.23 USD InStock
101779-284 101779-282
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Hazard: Flammable/Refrigerate


Formula: C₉H₁₉BO₃
Boiling Pt: 73°C
Density: 0.912
MDL Number: MFCD00192241
CAS Number: 61676-62-8
Order Now
Specification Test Results
Boiling point 73°
Density 0.912

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty