Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)pyridine ≥95%

Supplier: MATRIX SCIENTIFIC MS

007096-100G 007096-25G
101779-346EA 273.85 USD InStock
101779-346 101779-344
2-(Trifluoromethyl)pyridine ≥95%
2-(Trifluoromethyl)pyridine
Hazard: Flammable


Formula: C₆H₄F₃N
Boiling Pt: 139…141 °C
MDL Number: MFCD02183562
CAS Number: 368-48-9
Order Now
Specification Test Results
Boiling point 139-141°
ND20 1.418

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty