Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodobenzenesulphonyl chloride, Technical Grade

Supplier: MATRIX SCIENTIFIC MS

007147-5G 007147-1G
101779-414EA 623.08 USD InStock
101779-414 101779-412
2-Iodobenzenesulphonyl chloride, Technical Grade
2-Iodobenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₆H₄ClIO₂S
MW: 302.52
Melting Pt: 48…51 °C
MDL Number: MFCD00833336
CAS Number: 63059-29-0
Order Now
Specification Test Results
Melting point 48-51°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty