Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N1-Dimethylpentane-1,5-diamine

Supplier: MATRIX SCIENTIFIC MS

007229-1G 007229-5G
101779-522EA 223.08 USD InStock
101779-522 101779-524
N1,N1-Dimethylpentane-1,5-diamine
N1,N1-Dimethylpentane-1,5-diamine
Hazard: Corrosive/Stored under Argon


Formula: C₇H₁₈N₂
Boiling Pt: 50 °C ( 3 mmHg)
MDL Number: MFCD00047978
CAS Number: 3209-46-9
Order Now
Specification Test Results
Boiling point 50°/ 3mm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty