Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluorobenzylamine ≥95%

Supplier: MATRIX SCIENTIFIC MS

007237-25G 007237-5G
101779-530EA 103.08 USD InStock
101779-530 101779-528
2-Chloro-4-fluorobenzylamine ≥95%
2-Chloro-4-fluorobenzylamine
Hazard: Corrosive/Air Sensitive


Formula: C₇H₇ClFN
Boiling Pt: 94…95 °C (15 mmHg)
MDL Number: MFCD00042532
CAS Number: 15205-11-5
Order Now
Specification Test Results
Boiling point 94-95°/15m
ND20 1.535

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty