Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoroisobutyric acid ≥96%

Supplier: MATRIX SCIENTIFIC MS

007488-1G 007488-25G 007488-5G
101779-562EA 21.54 USD InStock
101779-562 101779-566 101779-564
2-Fluoroisobutyric acid ≥96%
2-Fluoroisobutyric acid
Hazard: Toxic/Corrosive/Refrigerate


Formula: C₄H₇FO₂
Boiling Pt: 57…58 °C (10 mmHg)
MDL Number: MFCD03452788
CAS Number: 63812-15-7
Order Now
Specification Test Results
Boiling point 57-58°/10m

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty