Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Diethylamino)pentyl alcohol ≥97%

Supplier: MATRIX SCIENTIFIC MS

007558-5G 007558-25G
101779-614EA 144.62 USD InStock
101779-614 101779-616
5-(Diethylamino)pentyl alcohol ≥97%
5-(Diethylamino)pentyl alcohol
Hazard: Corrosive


Formula: C₉H₂₁NO
MW: 159.27 g/mol
Boiling Pt: 131 °C
Density: 0.8842
MDL Number: MFCD00046024
CAS Number: 2683-57-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty