Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p,α,α,α-Tetrafluoroanisole ≥98%

Supplier: MATRIX SCIENTIFIC MS

008766-25G 008766-100G
101779-754EA 29.23 USD InStock
101779-754 101779-756
p,α,α,α-Tetrafluoroanisole ≥98%
p,α,α,α-Tetrafluoroanisole
Hazard: Flammable


Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 104…105 °C
Density: 1.323
MDL Number: MFCD00040835
CAS Number: 352-67-0
Order Now
Specification Test Results
Boiling point 104-105°
Density 1.323
ND20 1.394

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty