Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3,3,3-trifluoropropionate ≥97%

Supplier: MATRIX SCIENTIFIC MS

008937-5G 008937-1G
101779-798EA 90.77 USD InStock
101779-798 101779-796
Ethyl-3,3,3-trifluoropropionate ≥97%
Ethyl-3,3,3-trifluoropropionate
Hazard: Flammable/Irritant


Formula: C₅H₇F₃O₂
Boiling Pt: 50 °C (12 mmHg)
Melting Pt: 109 °C
MDL Number: MFCD01348113
CAS Number: 352-23-8
Order Now
Specification Test Results
Boiling point 50°/12mm
ND20 1.3912

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty