Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,2-Dibromo-5-fluorotoluene ≥98%

Supplier: MATRIX SCIENTIFIC MS

011284-25G 011284-100G
101780-084EA 106.15 USD InStock
101780-084 101780-086
α,2-Dibromo-5-fluorotoluene ≥98%
α,2-Dibromo-5-fluorotoluene
Hazard: Corrosive/Refrigerate/Lachrymator


Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 89…90 °C (1 mmHg)
Melting Pt: 33…36 °C
MDL Number: MFCD00042187
CAS Number: 112399-50-5
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Boiling point 89-90°/1mm
ND20 1.59

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty