Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-m-toluidine ≥99%

Supplier: MATRIX SCIENTIFIC MS

012514-25G 012514-100G
101780-106EA 23.08 USD InStock
101780-106 101780-108
6-Fluoro-m-toluidine ≥99%
6-Fluoro-m-toluidine
Hazard: Toxic/Irritant


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 88…90 °C (17 Torr)
MDL Number: MFCD00007654
CAS Number: 452-84-6
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty