Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anthranilonitrile ≥98%

Supplier: MATRIX SCIENTIFIC MS

012614-25G 012614-100G
101780-130EA 50.77 USD InStock
101780-130 101780-132
Anthranilonitrile ≥98%
Anthranilonitrile
TCA listed: T
Hazard: Toxic/Irritant


Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 268
Melting Pt: 47
MDL Number: MFCD00007631
CAS Number: 1885-29-6
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Boiling point 148-150°/2

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty