Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Undecafluoro-2-methyl-3-oxahexanoic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

018340-5G 018340-25G
101780-234EA 49.23 USD InStock
101780-234 101780-236
Undecafluoro-2-methyl-3-oxahexanoic acid ≥97%
Undecafluoro-2-methyl-3-oxahexanoic acid
Hazard: Corrosive/Irritant


Formula: C₆HF₁₁O₃
Boiling Pt: 186-188°C
MDL Number: MFCD00236734
CAS Number: 13252-13-6
Order Now
Specification Test Results
Boiling point 186-188°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty