Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluorobenzenesulphonyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

024303-25G 024303-5G
101780-358EA 83.08 USD InStock
101780-358 101780-356
4-Bromo-3-fluorobenzenesulphonyl chloride ≥97%
4-Bromo-3-fluorobenzenesulphonyl chloride
Hazard: Corrosive


Formula: C₆H₃BrClFO₂S
MW: 273.51
Boiling Pt: 96…97 °C (0.1 mmHg)
Density: 1.85
MDL Number: MFCD03094150
CAS Number: 351003-51-5
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Boiling point 96-97°/0.1
Density 1.85
ND20 1.577

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty