Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-5-methylpyrimidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

024837-5G 024837-25G
101780-454EA 67.69 USD InStock
101780-454 101780-456
4,6-Dichloro-5-methylpyrimidine ≥95%
4,6-Dichloro-5-methylpyrimidine
Hazard: Corrosive


Formula: C₅H₄Cl₂N₂
MW: 163.01
MDL Number: MFCD01646112
CAS Number: 4316-97-6
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty