Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%

Supplier: MATRIX SCIENTIFIC MS

032429-25G 032429-1G
101780-532EA 573.85 USD InStock
101780-532 101780-528
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
Hazard: Toxic/Irritant-Harmful/Refrigerate/Lachrymator


Formula: C₈H₃F₄NO
MW: 205.11
Boiling Pt: 53
Density: 1.418
MDL Number: MFCD00673069
CAS Number: 69922-27-6
UN: 2922
ADR: 8 III
Order Now
Specification Test Results
Boiling point 53°/2.5mm

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty