Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-ethylenediamine ≥95%

Supplier: MATRIX SCIENTIFIC MS

053731-25G 053731-100G
101780-648EA 38.46 USD InStock
101780-648 102833-180
N-Boc-ethylenediamine ≥95%
N-Boc-ethylenediamine
Hazard: Corrosive


Formula: C₇H₁₆N₂O₂
Boiling Pt: 72°C/0.1mm
Density: 1.016
MDL Number: MFCD00191871
CAS Number: 57260-73-8
Order Now
Specification Test Results
Boiling point 72°/0.1mm
Density 1.016

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty