Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butyl isothiocyanate ≥99%

Supplier: MATRIX SCIENTIFIC MS

056940-50G 056940-250G 056940-10G
101780-704EA 286.15 USD InStock
101780-704 101780-706 101780-702
1-Butyl isothiocyanate ≥99%
1-Butyl isothiocyanate
Hazard: Toxic/Corrosive/Refrigerate


Formula: C₅H₉NS
MW: 115.2 g/mol
Boiling Pt: 66
Density: 0.95
MDL Number: MFCD00004824
CAS Number: 592-82-5
UN: 1693
ADR: 6.1 II
Order Now
Specification Test Results
Boiling point 70-71°/35m
Density 0.95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty