Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclooctyl isothiocyanate ≥98%

Supplier: MATRIX SCIENTIFIC MS

056991-25G 056991-5G
101780-728EA 1044.62 USD InStock
101780-728 101780-726
Cyclooctyl isothiocyanate ≥98%
Cyclooctyl isothiocyanate
Hazard: Toxic/Corrosive


Formula: C₉H₁₅NS
Boiling Pt: 160 °C (22 mmHg)
Density: 1.01
MDL Number: MFCD00040882
CAS Number: 33522-04-2
Order Now
Specification Test Results
Boiling point 160°/22mm
Density 1.01

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty