Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TRPM7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB500-243SS NB500-243
102132-104EA 183.38 USD InStock
102132-104 102132-100
Anti-TRPM7 Rabbit Polyclonal Antibody
Antibodies
The TRPM7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRPM7. This antibody reacts with human, mouse (negative). The TRPM7 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: TRPM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse (Negative)

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty