Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Calcium-sensing R/CaSR Mouse Monoclonal Antibody [clone: HL 1499]

Supplier: Novus Biologicals
NB100-1830 NB100-1830SS
102106-110EA 503.38 USD InStock
102106-110 102106-122
Anti-Calcium-sensing R/CaSR Mouse Monoclonal Antibody [clone: HL 1499]
Antibodies
The Calcium-sensing R / CaSR Antibody (HL 1499) from Novus Biologicals is a mouse monoclonal antibody to Calcium-sensing R / CaSR. This antibody reacts with human, mouse, rat, rabbit. The Calcium-sensing R / CaSR Antibody (HL 1499) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunoassay.

Simple Western , Radioimmunoassay

Type: Primary
Antigen: Calcium-sensing R/CaSR
Clonality: Monoclonal
Clone: HL 1499
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty