Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 24 1-68]

Supplier: Novus Biologicals
NB500-211 NB500-211SS
102131-996EA 343.38 USD InStock
102131-996 102131-998
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 24 1-68]
Antibodies
The CHAF1B Antibody (SS 24 1-68) from Novus Biologicals is a mouse monoclonal antibody to CHAF1B. This antibody reacts with human, mouse, primate. The CHAF1B Antibody (SS 24 1-68) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: CHAF1B
Clonality: Monoclonal
Clone: SS 24 1-68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Primate

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty