Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-HIF1A Mouse Monoclonal Antibody [clone: HA111]

Supplier: Novus Biologicals
NB100-296 NB100-296SS
102108-394EA 417.23 USD InStock
102108-394 102108-406
Anti-HIF1A Mouse Monoclonal Antibody [clone: HA111]
Antibodies
Mouse Monoclonal HIF-1 alpha Antibody (HA111). Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty