Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ARNT/HIF-1 beta Mouse Monoclonal Antibody [clone: H1beta234]

Supplier: Novus Biologicals
NB100-124 NB100-124SS
102104-850EA 564.92 USD InStock
102104-850 102104-874
Anti-ARNT/HIF-1 beta Mouse Monoclonal Antibody [clone: H1beta234]
Antibodies
The ARNT / HIF-1 beta Antibody (H1beta234) from Novus Biologicals is a mouse monoclonal antibody to ARNT / HIF-1 beta. This antibody reacts with human, mouse, rat, bovine, ferret, primate, sheep. The ARNT / HIF-1 beta Antibody (H1beta234) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: ARNT/HIF-1 beta
Clonality: Monoclonal
Clone: H1beta234
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Bovine, Ferret, Primate, Sheep

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty