Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Sodium Potassium ATPase Beta 1 Mouse Monoclonal Antibody [clone: 464.8 (also known as 8A)]

Supplier: Novus Biologicals
NB300-147 NB300-147SS
102130-032EA 503.38 USD InStock
102130-032 102130-034
Anti-Sodium Potassium ATPase Beta 1 Mouse Monoclonal Antibody [clone: 464.8 (also known as 8A)]
Antibodies
The Sodium Potassium ATPase Beta 1 Antibody (464.8 (also known as 8A)) from Novus Biologicals is a mouse monoclonal antibody to Sodium Potassium ATPase Beta 1. This antibody reacts with human, rat, porcine, bovine, canine, hamster, rabbit, xenopus, mouse (negative). The Sodium Potassium ATPase Beta 1 Antibody (464.8 (also known as 8A)) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Sodium Potassium ATPase Beta 1
Clonality: Monoclonal
Clone: 464.8 (also known as 8A)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat, Porcine, Bovine, Canine, Hamster, Rabbit, Xenopus, Mouse (Negative)

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty