Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-STRO-1 Mouse Monoclonal Antibody [clone: STRO-1]

Supplier: Novus Biologicals
NBP1-48356 NBP1-48356SS
102178-598EA 503.38 USD InStock
102178-598 102178-610
Anti-STRO-1 Mouse Monoclonal Antibody [clone: STRO-1]
Antibodies
The STRO-1 Antibody (STRO-1) from Novus Biologicals is a mouse monoclonal antibody to STRO-1. This antibody reacts with human. The STRO-1 Antibody (STRO-1) has been validated for the following applications: Flow Cytometry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: STRO-1
Clonality: Monoclonal
Clone: STRO-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Lambda
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty