Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-beta-1,3-Glucuronyltransferase 1/B3GAT1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78982 NBP1-78982SS
102213-592EA 503.38 USD InStock
102213-592 102213-594
Anti-beta-1,3-Glucuronyltransferase 1/B3GAT1 Rabbit Polyclonal Antibody
Antibodies
The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-1,3-Glucuronyltransferase 1 / B3GAT1. This antibody reacts with human, mouse. The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:600

Type: Primary
Antigen: beta-1,3-Glucuronyltransferase 1/B3GAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty